Bulk and stoppering tray dryers, FreeZone®
Leverandør: LABCONCO
7806060 7806061 7948060 780601030 780601130 780601230 780601330 794801030 794801130 794801230 794801330 79480123031812
LBCP7806060EA 130475 NOK
LBCP7806060 LBCP7806061 LBCP7948060 LBCP780601030 LBCP780601130 LBCP780601230 LBCP780601330 LBCP794801030 LBCP794801130 LBCP794801230 LBCP794801330 LBCP79480123031812
Bulk and stoppering tray dryers, FreeZone®
Frysetørkere
The FreeZone® bulk and stoppering tray dryers have a stainless steel interior and epoxy coated, stainless steel exterior. Units include three probes for monitoring sample and shelf temperature. Microprocessor controlled temperature programming from −40 to +40 °C during ramping and holding in stoppering tray dryers, whereas in bulk dryers a microprocessor maintains the system temperature within 3 °C of the set point (maximum 60 °C). Units have an LCD display and audible/visual alarm indication for temperature and vacuum changes.
  • Compact design for moderate to large bulk loads or