Watertight pan cart
Leverandør: LABCONCO
8047500
LBCP8047500EA 0 NOK
LBCP8047500
Watertight pan cart
Traller
Watertight pan cart with durable 18-gauge welded steel frame has a glacier white epoxy-coated rust-resistant finish. Ideal for transporting cellware, cages, glassware, and more.
  • Watertight epoxy-coated pans are 5,1 cm (2") deep with removable rubber mats to provide a cushioned surface and eliminate sliding during transport
  • Cart features four 10,2 cm (4") diameter casters (two with toe locks) and two handles with ergonomic extruded rubber grips
  • Durable 18-gauge welded steel frame has a glacier white epoxy-coated rust-resistant finish.
  • Holds up to 181 kg (400