WaterPro® PS HPLC/UF Hybrid Polishing Station
Leverandør: LABCONCO
9000732
LBCP9000732EA 0 NOK
LBCP9000732
WaterPro® PS HPLC/UF Hybrid Polishing Station
Vannrensesystemer Ionebyttere
WaterPro PS Polishing Stations deliver Type I, 18.2 megohm-cm water from the dispensing valve or optional dispensing gun (refer to descriptions for models that include gun) at a typical rate of 1.8 liters per minute depending on feedwater flow rate and pressure, and installed purification modules.
The control panel includes a timed dispense feature that allows unattended dispensing of pure water without risk of overfilling. The dispenser shuts off automatically once user-set time has elapsed. Labconco has designed WaterPro PS Stations with the right combinations of purificationMore Product Information