Pads, D-Formalizer®
Leverandør: LEICA HISTOLOGY
38007530810
LEIH38007530810EA 2750 NOK
LEIH38007530810
Pads, D-Formalizer®
Benkepapir Benkepapir
These D-Formalizer® pads reduces formaldehyde exposure by absorbing and neutralising any excess formalin.
  • Highly durable
  • Can be used to line shelves
  • Highly contrasting blue surface
The durable blue surface of the pads provides contrast to even small pieces of tissue. The Pads are also useful for soaking up leakage and de-odorising vapors in other applications. They can be used to line shelves which hold specimen containers that may seep formaldehyde around the lid, or to line transport boxes where leaks may occur.