Microsystems™ Schott Gooseneck Light Guide

Leverandør: LEICA MICROSYSTEMS
31154202
LEIM31154202EA 6075 NOK
LEIM31154202
Microsystems™ Schott Gooseneck Light Guide
Lys Light Guides
High quality schott fiber optics for perfect light placement. Microsystems™ Schott Gooseneck Light Guide provides flex and stay lighting.

  • Precise positioning offers complete control of light placement
  • Gooseneck branches away from light for near shadow-free illumination
  • Easily adjustable, but holds its position
  • Length: 600 mm
  • Diameter : 4,5 mm

Leveringsinformasjon: Includes with two arms.
Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR