Stoppekraner, treveis, Czako
Leverandør: Lenz Laborglas GmbH & CO.KG
02631804
LENZ02631804EA 2475 NOK
LENZ02631804
Stoppekraner, treveis, Czako
Haner
Stopcocks, made of DURAN® borosilicate glass.
  • Good temperature resistance
  • Good chemical resistance
With PTFE plug and screwthread retaining device.
Sertifiseringer: Leakage rate according to DIN 12540-1.