Destillasjonsoppsats for kortveisdestillasjon
Leverandør: Lenz Laborglas GmbH & CO.KG
05058001
LENZ05058001EA 3250 NOK
LENZ05058001
Destillasjonsoppsats for kortveisdestillasjon
Destillasjonsutstyr Broer
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Short condensate path due to compact design of condenser and vacuum connection, with two cones NS 14/23, one socket NS 14/23 and three DIN threads GL 14.