Adaptere, bøyde
Leverandør: Lenz Laborglas GmbH & CO.KG
05313044
LENZ05313044EA 1550 NOK
LENZ05313044
Adaptere, bøyde
Destillasjonsutstyr Destillasjonsadaptere, flere halser
DURAN® Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Connector, bent, with cone and lateral socket NS 29/32, with socket NS 14/23 (for thermometer or air leak tube).