Tørkerør, U-formede
Leverandør: Lenz Laborglas GmbH & CO.KG
05339010 05339013 05339018
LENZ05339010EA 565 NOK
LENZ05339010 LENZ05339013 LENZ05339018
Tørkerør, U-formede
Rør Tørkerør
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Drying tubes, U-shaped, side tubes with olives (Ext.Ø: 6 mm).