Uttrekker for faste stoffer
Leverandør: Lenz Laborglas GmbH & CO.KG
05431137 05431131 05431149
533-2151EA 2475 NOK
533-2151 LENZ05431131 LENZ05431149
Uttrekker for faste stoffer
Ekstraktorer Soxhlet- og Soxhlet-typeuttrekkere
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
For glass inserts for solids.