Stirrer with paddles
Leverandør: Lenz Laborglas GmbH & CO.KG
06308001 06308003 06308005 06308007 06308009
LENZ06308001EA 2500 NOK
LENZ06308001 441-0049 LENZ06308005 LENZ06308007 LENZ06308009
Stirrer with paddles
Omrørere Omrører-utstyr
DURAN®, borosilicate glass shaft, PFTE paddles with ground and polished shaft.
NB: * stirring Ø when blades extended