Stirrers with PTFE blade
Leverandør: Lenz Laborglas GmbH & CO.KG
06315001 06315003 06315005 06315007 06315008 06315011 06315015 06315031 06315035 06315037
LENZ06315001EA 1900 NOK
LENZ06315001 LENZ06315003 LENZ06315005 LENZ06315007 LENZ06315008 LENZ06315011 LENZ06315015 LENZ06315031 LENZ06315035 LENZ06315037
Stirrers with PTFE blade
Omrørere Omrører-utstyr
Stirrer made of precision calibrated glass tube and shaft made of DURAN® tubing with PTFE blade.
  • With ground and polished bearing surface
  • With snap-fastener for replacement of PTFE blades