Perfection and Performance Stainless-steel Freezers for Professional-use, FFPSvh & FFPSvg
Leverandør: Liebherr
FFPSVG6501
LIEBFFPSVG6501EA 0 NOK
LIEBFFPSVG6501
Perfection and Performance Stainless-steel Freezers for Professional-use, FFPSvh & FFPSvg
Frysere Frysere, generelt bruk
High-volume general-purpose freezers with forced air cooling. Stainless-steel exterior and interior. Temperature stability in accordance with EN 16825.
  • Energy-efficiency optimised appliances for lower running costs and help to meet user sustainability targets
  • Equipped with 150 mm legs, allows cleaning underneath
  • Mechanical door locks for basic content security
  • Forced-air cooling system with energy-efficient hot gas defrost
  • Power failure alarm when mains power returns
  • Wire shelves with 60 kg shelf load
Choose between two execution options to suitMore Product Information