FNe 5207 Pure Freestanding freezer, with NoFrost
Leverandør: Liebherr
FNE5207
LIEBFNE5207EA 0 NOK
LIEBFNE5207
FNe 5207 Pure Freestanding freezer, with NoFrost
Frysere Frysere, generelt bruk
NoFrost protects the freezer compartment from unwanted icing, which consumes a lot of energy and can be expensive. NoFrost means no more tedious and time-consuming defrosting of the freezer compartment, more time for other things and saving money.
  • Frost protect
  • Drawer with comfortable tilting
  • Energy saver
  • Power failure alarm
  • Door alarm
  • Vario space
  • Closing assistance
  • Adjustable feet
  • Replaceable door seal
  • Dishwasher-safe features
The touch display enables easy and intuitive operation. Simple, classic and high-quality with its shiny white-painted surface.More Product Information