Reach-In Refrigerator with Bottom Compressor, FRFCvg
Leverandør: Liebherr
FRFCVG5501 FRFCVG6511 FRFCVG5511
LIEBFRFCVG5501EA 0 NOK
LIEBFRFCVG5501 LIEBFRFCVG6511 LIEBFRFCVG5511
Reach-In Refrigerator with Bottom Compressor, FRFCvg
Kjøleskap
This FRFCvg Reach-In refrigerator with bottom compressor save time and money. In case of an uncontrolled temperature rise, if a door has been left open or there were any other problems, the interior light of the appliance will start flashing.
  • Environmentally friendly refrigerants
  • Efficient refrigeration system
  • Interchangeable door hinge
  • Display lock
  • Mechanical lock
  • Easy to clean interior
  • Grid shelf with pull-out stop
  • Digital temperature display
  • Reversible door hinges (left/right)
This refrigerators for the catering industry are generally designed forMore Product Information