Pharmaceutical Underbench Perfection Refrigerators Compliant with DIN 13277, HMTvh
Leverandør: Liebherr
HMTVH1501 HMTVH1501_H63 HMTVH1511 HMTVH1511_H63
LIEBHMTVH1501EA 0 NOK
LIEBHMTVH1501 LIEBHMTVH1501_H63 LIEBHMTVH1511 LIEBHMTVH1511_H63
Pharmaceutical Underbench Perfection Refrigerators Compliant with DIN 13277, HMTvh
Kjøleskap
Ultra-efficient pharmaceutical refrigerators have been designed for the particular requirements with their dynamic cooling system, highly effective insulation, and precise temperature control, they guarantee reliable storage of sensitive substances and preparations.
  • Integrated data logger and USB interface for reading the data logger
  • PT1000 product sensor and access port for external probes
  • Min./max. temperature log shows whether the storage temperatures were within the tolerance limits
  • Insulated and glass door models with vertical LED lighting, self-