Spark-free Performance laboratory freezers with ATEX certificate, SFFfg
Leverandør: Liebherr
SFFFG4001 SFFFG5501 SFFFG4001_740 SFFFG5501_740
LIEBSFFFG4001EA 33625 NOK
LIEBSFFFG4001 LIEBSFFFG5501 471-1343 LIEBSFFFG5501_740
Spark-free Performance laboratory freezers with ATEX certificate, SFFfg
Frysere Frysere, generelt bruk
Freezers with a spark-free interior specifically for storing explosive and highly flammable substances in laboratories and in the chemical industry.
  • Alarm relay, alarm test function and custom alarm settings possible
  • Power failure alarm when mains power returns
  • Min./max. temperature log, self-closing door at <90° and mechanical lock provided
  • Optional LAN/WiFi, Local API (optional via SmartModule)
  • SmartModule can be retrofitted
  • Antimicrobial handle with opening mechanism
  • Super-efficient energy consumption
The plastic inner liner is made ofMore Product Information