Performance and Perfection Laboratory Freezers with Stainless Steel Inner Liner, SFPvh and SFPvg
Leverandør: Liebherr
SFPVG6501
LIEBSFPVG6501EA 0 NOK
LIEBSFPVG6501
Performance and Perfection Laboratory Freezers with Stainless Steel Inner Liner, SFPvh and SFPvg
Frysere Frysere, generelt bruk
High-volume freezers with forced air cooling. White steel exterior and stainless-steel interior. The intuitive user interfaces provide useful extended multi-functionality, e.g. a maintenance timer, and the LED interior lighting ensures good visibility of the contents of these large appliances. Advanced connectivity options to suit varying requirements for simplified data storage, temperature control and alarm signals.
  • Extended range of models for more choice to fulfil specific user requirements
  • Hygienic and long-lasting stainless-steel interior with a 12