Spark-free Performance laboratory refrigerators with ATEX certificate, SRFfg
Leverandør: Liebherr
SRFFG5501 SRFFG3501 SRFFG4001
LIEBSRFFG5501EA 35950 NOK
LIEBSRFFG5501 471-1340 LIEBSRFFG4001
Spark-free Performance laboratory refrigerators with ATEX certificate, SRFfg
Kjøleskap
The refrigerators with spark-free interior are easy to clean and the robust glass shelves are simple to remove at 90° door opening. They are super-efficient, utilising the energy-savings potential of every component.
  • Alarm relay, alarm test function and custom alarm settings possible
  • Min./max. temperature log, self-closing door at <90° and mechanical lock provided
  • Retrofittable optional LAN/WiFi, Local API (optional via SmartModule)
  • SmartModule can be retrofitted
  • Antimicrobial handle with opening mechanism
  • Super-efficient energy consumption and forced-