Perfection and Performance Laboratory Refrigerators with Stainless Steel Inner Liner, SRPvh & SRPvg
Leverandør: Liebherr
SRPVG6501
LIEBSRPVG6501EA 72125 NOK
LIEBSRPVG6501
Perfection and Performance Laboratory Refrigerators with Stainless Steel Inner Liner, SRPvh & SRPvg
Kjøleskap
Glass/steel door high-volume refrigerators with forced air cooling. White steel exterior and stainless-steel interior. Choose between two temperature stability options to suit differing lab requirements. The intuitive user interfaces provide useful extended multi-functionality, e.g. a maintenance timer, and the LED interior lighting ensures good visibility of the contents of these large appliances. Advanced connectivity options to suit varying requirements for simplified data storage, temperature control and alarm signals. These advanced lab fridges also feature a +2 °C safety device toMore Product Information