Slide box
Leverandør: LIEDER JOHANNES
PK25
LIEDPK25EA 75 NOK
LIEDPK25
Slide box
Objektglassbokser
This slide box is designed to facilitate the transportation of 25 microscope slides.
  • Transparent cover for easy viewing
  • Robust design and stackable construction
  • Serrated retainer strips to maintain slides properly