PAR test agar
Leverandør: LIOFILCHEM
11200
LIOF11200EA 1225 NOK
LIOF11200
PAR test agar
Mikrobiologiske medier Media, preparerte for mikrobiologi
Medium with spores of Geobacillus stearothermophilus subsp. calidolactis for residual antibiotic detection in biological liquids.
  • For milk, blood and urine