Culture Media, Ready to Use, in Bags
Leverandør: LIOFILCHEM
499070
LIOF499070EA 1900 NOK
LIOF499070
Culture Media, Ready to Use, in Bags
Mikrobiologiske medier Media, dehydrert for mikrobiologi
Microbiology culture media bags, are pre-prepared, sterile liquid culture media contained in plastic bags. They are used in microbiology laboratories for various applications, including food and pharmaceutical microbiology.
  • Time saving: Since the media is pre-prepared, there is no preparation time required. You can start using the media immediately upon arrival in the lab
  • Reduced risk of contamination: Pre-sterilised media reduces the risk of contamination compared to manually prepared media
  • Batch consistency: Ordering large batches of RTU media can reduce the number of performance tests required, ensuring consistency between experiments
These bags are designed to be connected to a gravimetric dilutor, simplifying the process of weighing and diluting samples.