Dehydrated Culture Media, Columbia Agar Base
Leverandør: LIOFILCHEM
6100135
LIOF6100135EA 10450 NOK
LIOF6100135
Dehydrated Culture Media, Columbia Agar Base
Mikrobiologiske medier Media, dehydrert for mikrobiologi
Columbia agar base, enriched with sterile sheep blood (5%), is suitable for isolation and growth of fastidious microorganisms such as streptococci, staphylococci, pneumococci and listeriae from clinical samples.