Latex Kit for Antibody Determination
Leverandør: LIOFILCHEM
96117
LIOF96117EA 0 NOK
LIOF96117
Latex Kit for Antibody Determination
Mikrobiologiske testsystemer Antigen-antistoff tester Ag-AK, Latex
This kit is for qualitative and semi-quantitative determination of Treponema pallidum antibodies.