LiChrospher®, HPLC Columns
Leverandør: Avantor
LISP60-5-150AEA 9900 NOK
LISP60-5-150A LISP60-5-250A LISPCN-5-250A LISPOH-5-250A LISP100-5-150A LISP100-5-250A LISP60-5-125AF HICHLISP100-5100AF HICHLISP100-5100AM HICHLISP100-5100AS LISP100-5-10C5 LISPOH-5-100AF LISPOH-5-10C5 LISPOH-5-10CM5 LISPOH-5-125AF LISPOH-5-150AM LISPOH-5-250AF LISPOH-5-250AM LISP60-5-10CP3 LISP60-5-250P LISPOH510C5SK LISPOH-5-10CE5 LISPOH-5-250P
LiChrospher®, HPLC Columns
Kromatografikolonner
LiChrospher® is a silica available in both endcapped and non-endcapped C8 and C18 phases.
  • Spherical porous silica
  • Base deactivated LiChrospher® RP-select B
  • High efficiency packed columns
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat ISO 14001:2015 (530840 UM15)