Turbidity meters, TurbiDirect
Leverandør: Lovibond
194000
LOVI194000EA 25275 NOK
LOVI194000
Turbidity meters, TurbiDirect
Turbiditetsmålere
The TurbiDirect is an autoranging turbidity meter designed for the laboratory or portable use.
  • Real time integrated clock
  • Waterproof sample chamber
  • Large graphic display for results easy to interpret
Sertifiseringer: Infrared light source and 90° detection angle conform to EN ISO 7027.
Leveringsinformasjon: Supplied in carrying case with turbidity standard (0,1 ; 20 ; 200 ; 800 NTU), seven AA batteries, NiMH rechargeable 1100 mAh, mains adapter, PC cable with RS232 interface, four sample vials with lids, guarantee sheet, certificate of compliance and instructions manual.