Test kits, amine, Comparator 2000+
Leverandør: Lovibond
412740
LOVI412740EA 10875 NOK
LOVI412740
Test kits, amine, Comparator 2000+
Miljø testkit
Comparator 2000+ is a versatile modular system for testing water.
  • Stable and accurate
  • Robust, compact and portable
  • Prism enabling colour matching
  • Diffuser plate is used to measure the turbidity
  • Longer path length cells be used for lower concentration analysis
Leveringsinformasjon: Test kit is supplied in a robust carrying case containing Comparator 2000+, amines comparator disc, extraction tube for result measurement, filter funnel and filter papers, 100 ml amber glass bottle, tablet reagents, crushing/stirring rod and instruction sheet.