Test kits, nitrite, Comparator 2000+
Leverandør: Lovibond
502371
LOVI502371EA 465 NOK
LOVI502371
Test kits, nitrite, Comparator 2000+
Miljø testkit
Comparator 2000+ is a versatile modular system for testing water.
  • Stable and accurate
  • Robust, compact and portable
  • Diffuser plate is used to measure the turbidity
  • Longer path length cells be used for lower concentration analysis
The integrated prism brings the glass standards of the test discs and the coloured sample into the same field of view.