Dipslide incubator, DI 10
Leverandør: Lovibond
56B000701
LOVI56B000701EA 9375 NOK
LOVI56B000701
Dipslide incubator, DI 10
Inkubatorer Inkubatorer, bordmodell
The DI 10 incubator is designed for reliable incubation of bacteriological slides, on-site, in a laboratory or even while mobile in a car or van.
  • Robust design
  • Holds up to 12 dipslides or 10 quanti-discs
  • With 1×16 bit backlit LCD display
  • Excellent temperature stability
  • Programmable incubation period setting
The incubation period and the incubation temperature should be the same each time the test is performed so that bacteria growth is controlled. This allows for week by week comparisons to be made, high counts easier to identify. Dipslides areMore Product Information
Bestill nå
SPESIFIKASJONER
Modell DI 10
Environmentally Preferable
Vekt 1,7 kg