VWR®, DNA extraction Kit for Real-time PCR food samples analysis
Leverandør: VWR Chemicals
M04018BFEA 2250 NOK
M04018BF
VWR®, DNA extraction Kit for Real-time PCR food samples analysis
Nukleinsyre reagenser Nukleinsyrerensingsreagenser og -kit
Rapid method for DNA extraction and purification from all types of microorganisms in a variety of matrices for molecular biology.
  • Rapid method for DNA extraction
  • 100 samples for VWR® Real-time PCR food samples kit