Tris-(hydroksymetyl)aminometan ≥99.8%, Proteomics Grade

Leverandør: VWR Chemicals
Warning

M151-1KGEA 3625 NOK
M151-1KG
Tris-(hydroksymetyl)aminometan ≥99.8%, Proteomics Grade
Tris-(hydroksymetyl)aminometan

Reagent for stabilisation of buffer formulations and electrophoresis of biological molecules.

Formel: NH₂C(CH₂OH)₃
Kokepunkt: 357 °C (1013 hPa)
Smeltepunkt: 169…172 °C
Tetthet: 1,328 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004679
CAS-nummer: 77-86-1
EINECS: 201-064-4
Merck Indeks: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Abs.@280 nm (1M, Water) 0.05
Arsenic <0.0005%
Copper <0.0001%
DNase NONE
Insolubles 0.005 %
Iron <0.0001%
Lead <0.0001%
Loss on Drying 1.0%
Magnesium 0.0005%
Protease NONE
Purity 99.8%
RNase NONE
Solubility (10%, Water) PASS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR