VWR®, Bovine aprotinin (from Lung)

Leverandør: VWR Chemicals

M167-10MGEA 3200 NOK
M167-10MG
VWR®, Bovine aprotinin (from Lung)
Proteiner

A competitive and reversible serine protease inhibitor of chymotrypsin, trypsin, kallikrein and plasmin. Not an inhibitor of Factor Xa and thrombin.


Activity (KIU/mg)      ≤4500


Purity (HPLC)      Pass

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR