VWR®, Bradford protein assay, proteomics grade
Leverandør: VWR Chemicals
M172-1LEA 3600 NOK
M172-1L
VWR®, Bradford protein assay, proteomics grade
Assay Proteinanalyser
The Bradford protein assay is the preferred colorimetric assay for quantifying total protein concentration. Based upon complex formation between basic and aromatic amino acid residues with Coomassie® Brilliant Blue G-250 dye, the Bradford method is easier, faster, and more sensitive than the Lowry method.
  • Sensitive, fast and consistent
  • Standard protocol for measuring concentrations from 10 to 100 μg
  • Microassay protocol for measuring concentrations from 1 to 10 μg
Leveringsinformasjon: Each kit includes Bradford Reagent (1 L) and bovine albumin standard, 0,5 mg/ml (1,5 ml). Bradford Reagent and BSA standard solution are also available separately.