Metanol absolute acetone-free

Leverandør: Honeywell Chemicals
Danger

Synonymer: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

M1775-1GA
M1775-1GAEA 1725 NOK
M1775-1GA
Metanol absolute acetone-free
Metanol
Formel: CH₃OH
MW: 32.04 g/mol
Kokepunkt: 64…65 °C (1.013 hPa)
Smeltepunkt: –98 °C
Tetthet: 0.79 g/cm³ (20 °C)
Flash punkt: 11 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004595
CAS-nummer: 67-56-1
EINECS: 200-659-6
FN: 1230
ADR: 3 II
REACH: 01-2119433307-44
Merck Indeks: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Vapor Pressure 410 mmHg ( 50 °C)
Appearance complying
Autoignition Temperature 725 °F
Refractive Index n20/D 1.329 (lit.)
Explosion Limit 36 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR