VWR® Separating Gel Buffer, 4X
Leverandør: VWR Chemicals
M197-500MLEA 6500 NOK
M197-500ML
VWR® Separating Gel Buffer, 4X
Buffere Buffere, elektroforese Buffere, separasjon
4X concentrated buffer solution (1,5 M Tris, pH 8,8 and 0,4% SDS). Used for preparation of standard acrylamide:bis-acrylamide gels as well as preparation of gels using PAGE-PLUS™ acrylamide solution.