Natriumperoksid ≥93.0%, granules, AR® ACS, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials
Danger

Synonymer: di-Sodium peroxide

MACR7864-04EA 7650 NOK
MACR7864-04
Natriumperoksid ≥93.0%, granules, AR® ACS, Macron Fine Chemicals™
Natriumperoksid
Formel: Na₂O₂
Smeltepunkt: 660 °C
Tetthet: 2,805 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003497
CAS-nummer: 1313-60-6
EINECS: 215-209-4
FN: 1504
ADR: 5.1,I
Merck Indeks: 13,08728

Bestill nå

Spesifikasjon Testresultater

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (Na₂O₂) (by KMnO₄ titrn) ≥93.0 %
ACS - Chloride (Cl) ≤0.002 %
ACS - Heavy Metals (as Pb) ≤0.002 %
ACS - Iron (Fe) ≤0.005 %
ACS - Phosphate (PO₄) ≤0.0005 %
ACS - Sulfate (SO₄) ≤0.001 %
Appearance (white to pale yellow, hygroscopic granules)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR