PCR clean-up, PCR normalizer kit, Axygen® AxyPrep™ Mag
Leverandør: Corning
MAG-PCR-NM-5
MAG-PCR-NM-5EA 4575 NOK
MAG-PCR-NM-5
PCR clean-up, PCR normalizer kit, Axygen® AxyPrep™ Mag
Nukleinsyre reagenser Nukleinsyrerensingsreagenser og -kit
The AxyPrep™ Mag PCR Normalizer Kit utilizes a paramagnetic bead-based purification system for PCR clean up.
  • Dual action: Normalization and post PCR clean-up enables significant reduction in processing and faster turnaround times
  • Scalable: Tube, 96-, and 384-well plate formats
  • Manual and automation-compatible protocol
  • Compatible with several downstream applications
Axygen® AxyPrep™ Mag PCR Normalizer Kit utilises a paramagnetic bead-based purification system for PCR clean up. AxyPrep™ Mag PCR Normalizer beads have limited binding surface, therefore by limiting the amount of beads added in a given purification reaction, pre-defined amounts of DNA can be isolated based on user need. PCR normalisation is accomplished during this purification process so that additional DNA quantification and dilution are not necessary.
Time, labour and reagent cost are greatly saved with the AxyPrep™ Mag normalisation purification chemistry and thus efficiently streamlining genomics downstream applications. The protocol mainly consists of binding, washing and elution steps, and can be performed directly in the thermal cycling plate and requires no centrifugation or filtration. The process can be automated for high throughput applications.
Bestill nå
SPESIFIKASJONER
Beskrivelse Axygen® AxyPrep™ Mag PCR Normalizer Kit
Environmentally Preferable
Applikasjon
Target molecule DNA
Refine target molecule Amplified/modified DNA
Sample type PCR and other enzymatic reactions
Method / Format Magnetic beads
Prep size Scalable