Test mixtures for gas chromatography
Leverandør: MACHEREY-NAGEL
722307 722310 722311 722316 722317 722371 722372 722320
MANA722307EA 1975 NOK
MANA722307 MANA722310 MANA722311 MANA722316 MANA722317 MANA722371 MANA722372 MANA722320
Test mixtures for gas chromatography
Standarder kromatografi standarder
These test mixtures are used for controlling the performance of fused silica capillary columns and the GC system.