Test strips, qualitative, sulphite
Leverandør: MACHEREY-NAGEL
90763
MANA90763EA 1175 NOK
MANA90763
Test strips, qualitative, sulphite
Miljø testkit
The test paper allows the quick and easy detection of sulphite and sulphur dioxide. In food analysis laboratories it is used to detect sulphite in meat products.
  • Colour changes: White to pink-red
In medical diagnostic it is used as a quick test on sulphite oxidase deficiency.
Forpakning: Pack of 100 strips 20×70 mm.