Luminometer, BioFix® Lumi-10
Leverandør: MACHEREY-NAGEL
940008
MANA940008EA 188050 NOK
MANA940008
Luminometer, BioFix® Lumi-10
Overvåkningssystemer Hygieneovervåkningssystemer
BioFix® Lumi-10 is a portable luminometer for the measurement of biological and chemical luminescence reactions with relatively constant light emission.
  • Mobile test system with a high sensitive detector
  • High-resolution graphic display
  • Data memory for up to 2000 test results
  • Variable measurement times to obtain optimum results
  • Serial RS232 interface for data transfer to PC
This device is ideal for environmental analysis/ecological toxicology: acute luminous bacteria tests, hygiene monitoring: ATP and biomass analysis and molecular biological andMore Product Information