HistoBond® Adhesive Microscope Slides with Circles
Leverandør: MARIENFELD
0810471 0810461
MARI0810471EA 1125 NOK
MARI0810471 MARI0810461
HistoBond® Adhesive Microscope Slides with Circles
Objektglass Adhesjons objektglass
For use in cytocentrifuges we offer our adhesive and positively charged HistoBond®+ slides with white rings. These serve as a microscope help to find the centrifuged cells more easily.
  • Made of soda lime glass of third hydrolytic class
  • Dimensions: ~76×26 mm
  • Thickness: ~ 1 mm
  • With white marking area of ~20 mm on one side
  • Pre-cleaned and ready for use
  • Autoclavable
For applications according to EU regulation 2017/746 on invitro diagnostic medical devices (IVDR), with CE-mark, recommended best before date and batch number for comprehensive information andMore Product Information