Slide folder, 'Munich'
Leverandør: MARIENFELD
6001313
MARI6001313EA 2225 NOK
MARI6001313
Slide folder, 'Munich'
Objektglassholdere
Cardboard base with three compartments (78×220 mm).
  • Each compartment can accomodate up to eight slides
  • Hinged cover with three windows (56×200 mm)