Swivel Roller Mixers, RM5F
Supplier: MARIENFELD
7202038
MARI7202038EA 42125 NOK
MARI7202038
Swivel Roller Mixers, RM5F
Roller Culture Apparatus
Allows the thorough mixing of samples and cell suspensions by wobbling and rolling motions.
  • Wobbling and rolling motions
  • Five rollers made of PVC, each with a length of 334 mm
  • Rollers Ø: 32 mm
  • EU plug
Order Now