Disposal box for blades, SafeBox
Leverandør: MARTOR ARGENTAX
108000.00
MARO108000.00EA 180 NOK
MARO108000.00
Disposal box for blades, SafeBox
Bokser Avfallsbeholdere
This box is designed for the safe deposition of 'used' snap-off blade segments, blade ends, straight/hook/industrial cutter/deburring scalpel and hobby crafts blades.
Absolutely safe and inaccessible to fingers.