Scalpel Blades, No.11
Leverandør: MARTOR ARGENTAX
11.50
MARO11.50EA 175 NOK
MARO11.50
Scalpel Blades, No.11
Skalpellblad og skaft Skalpellblad
High-quality blade made of steel. It is very sharp and has one usable cutting edge.
  • Two-sided ground-edge
  • Aingle bevel edge-ground blade facets
  • 1-cutting edge blade
Material thickness: 0,40 mm
Blade weight: 0,4 g
The blade can be used in the knives: Grafix Scalpel small, Trimmex Cuttograf.
Forpakning: Packaging unit (PU) 10/magazine (size 66×25×18 mm, weight 16,6 g). Outer pack quantity of 10 magazines (size 188×69×28 mm, weight 190 g).