Storage tubes, MATRIX™
Leverandør: THERMO MATRIX TECHNOLOGIES
4147 4148 4140 4170 4273 4247 4248 4249 4250 4252 4253 4271 4272 4274
MATR4147EA 7850 NOK
MATR4147 MATR4148 MATR4140 MATR4170 MATR4273 MATR4247 MATR4248 MATR4249 MATR4250 MATR4252 MATR4253 MATR4271 MATR4272 MATR4274
Storage tubes, MATRIX™
Rør Reagensrør
PP, translucent
  • Non sterile or sterile available
  • Tubes are 100% leak tested to ensure sample integrity and security
  • Tubes are validated to be free of DNAse, RNAse, endotoxins and cytotoxins
Store valuable samples where 2D tracking is not required with high-density Thermo Scientific™ Matrix Blank and Alphanumeric Storage Tubes, offered in manual and automation-friendly racks. Visual identification of samples can be made using the alphanumeric code on the bottom of each tube, and the tubes are compatible with the full line of seals and capping equipment.More Product Information