Robotic tips, Matrix
Leverandør: THERMO MATRIX TECHNOLOGIES
5361
MATR5361EA 20125 NOK
MATR5361
Robotic tips, Matrix
Pipettespisser
These automation tips are compatible with 384 format Beckman instruments, PerkinElmer and FLIPR instruments using CCS Type A pipetting heads and FLIPR Type B Liberty systems. All of these tips are validated on their compatible instruments to ensure the highest quality for pipetting in automated laboratories.
  • Performance validation on compatible instruments includes 1536 target straightness testing, leak testing, length, weight and wall thickness testing, and pipetting head seal force testing
  • Design includes molded conductive rack and snap plate to hold