Pipettespisser, Automatpipettespisser, Matrix® D.A.R.Ts®
Leverandør: THERMO MATRIX TECHNOLOGIES
5506 5301 5302 5311 5312 5316 5317 5318 5321 5322 5326 5327 5328 5416 5417 5418 5507 5516 5517 5518 5526 5527 5528 5536 5537 5538 5546 5547 5548 5586 5587 5588
MATR5506EA 5750 NOK
MATR5506 MATR5301 MATR5302 MATR5311 MATR5312 MATR5316 MATR5317 MATR5318 MATR5321 MATR5322 MATR5326 MATR5327 MATR5328 MATR5416 MATR5417 MATR5418 MATR5507 MATR5516 MATR5517 MATR5518 MATR5526 MATR5527 MATR5528 MATR5536 MATR5537 MATR5538 MATR5546 MATR5547 MATR5548 MATR5586 MATR5587 MATR5588
Pipettespisser, Automatpipettespisser, Matrix® D.A.R.Ts®
Pipettespisser
D.A.R.T’S™ (Disposable Automation Research Tips) have been designed for the ultimate performance in automated pipetting.
  • Small internal orifice provides small droplet formation resulting in optimal accuracy and precision
  • Entire assembly can be quickly removed and replaced eliminating extra handling preventing damage or contamination
  • 100% inspection ensures superior quality
  • Flexibility to choose tips for any application
  • Available in standard, filtered, wide bore and extended length
Unlike conventional automation tips, D.A.R.T.’S are in disposable rigidMore Product Information