Manual microplate heat sealer, ALPS™ 30
Leverandør: THERMO MATRIX TECHNOLOGIES
AB-0400
MATRAB-0400EA 62200 NOK
MATRAB-0400
Manual microplate heat sealer, ALPS™ 30
Varmeforseglere Varmeforsegler for mikroplater
Use of the ALPS™ 30 manual heat sealer protects samples from evaporation and contamination when performing PCR or sample storage. Perfect for sealing small numbers of plates in low to medium throughput applications, the ALPS™ 30 unit is small enough to keep on the bench and not take up a lot of space. Its simple, ergonomic manual operation provides quick, efficient sealing.
  • Compatible with virtually any height microplate or block, including PCR plates
  • Consistent heat seal provides dependable protection for PCR or sample storage plates
  • Prevents